Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549508

Catalog No. PA549508

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable transcription factor involved in neuronal differentiation and/or phenotypic maintece.
Specifications

Specifications

ZFP2
Polyclonal
PBS with 0.09% sodium azide
Q6ZN57
Zinc finger protein 2 homolog, Zfp-2, Zinc finger protein 751, ZFP2, ZNF751
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80108
Western Blot
Unconjugated
ZFP2
Liquid
ZFP2
KLH conjugated synthetic peptide between 379-411 amino acids from human ZFP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit