Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zfp275 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19131920UL

 View more versions of this product

Catalog No. NBP19131920


Add to cart

Description

Description

Zfp275 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

Zfp275
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
Zfp275
The specific Immunogen is proprietary information. Peptide sequence LAEHQRIHSGAKPYGCPHCGKLFRRSSELTKHRRIHTGEKPYECNQCGKA.
Immunogen affinity purified
RUO
Primary
27081
Western Blot
Western Blot 1:1000
NP_113682
DXBay20, zinc finger protein 275
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit