Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP30 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548578

Catalog No. PA548578

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZFP30
Polyclonal
PBS with 0.09% sodium azide
Q9Y2G7
Zinc finger protein 30 homolog, Zfp-30, Zinc finger protein 745, ZFP30, KIAA0961, ZNF745
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22835
Western Blot
Unconjugated
ZFP30
Liquid
ZFP30
KLH conjugated synthetic peptide between 65-94 amino acids from the N-terminal region of human ZFP30.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit