Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP36L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc ZFP36L2201AP
Description
ZFP36L2 Polyclonal antibody specifically detects ZFP36L2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
ZFP36L2 | |
Polyclonal | |
Unconjugated | |
ZFP36L2 | |
Brf2; Butyrate response factor 2; EGF-response factor 2; ERF2; ERF-2; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36L2; mRNA decay activator protein ZFP36L2; Protein TIS11D; RNF162C; TIS11D; TISD; TPA-induced sequence 11d; ZFP36 ring finger protein like 2; ZFP36 ring finger protein-like 2; ZFP36L2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2 | |
Synthetic peptide taken within amino acid reigon 350-400 on human Zinc finger protein 36, C3H1 type-like 2. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P23949, P47974 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12193, 298765, 678 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction