Learn More
ZFP37 Mouse anti-Human, Clone: OTI3H4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810229
Catalog No. 50-166-9957
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZFP37 may be involved in transcriptional regulation.
Specifications
Specifications
ZFP37
Monoclonal
Unconjugated
ZFP37
Lyophilized
ZFP37
Full length human recombit protein of human ZFP37 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI3H4
PBS with 8% trehalose and no preservative; pH 7.3
Q9Y6Q3
ZNF906, zfp-37
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7539