Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP64 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$337.75 - $627.50
Specifications
Antigen | ZFP64 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ZFP64 Polyclonal specifically detects ZFP64 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZFP64 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
55734 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SFDTKQPSNLSKHMKKFHGDMVKTEALERKDTGRQSSRQVAKLDAKKSFHCDICDASFMREDSLRSHKRQHSEYSESKNSDVTVLQFQIEPS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
dJ548G19.1, dJ831D17.1, FLJ10734, FLJ12628, Zfp-64, Zinc finger protein 338MGC940, zinc finger protein 64, zinc finger protein 64 homolog (mouse), ZNF338 | |
ZFP64 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title