Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP69 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555696
Description
Immunogen sequence: EEPWMAEKEG PGDPSSDLKS KIETIESTAK STISQERLYH GIMMESFMRD DIIYSTLRKV STYDDVLERH QETC Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.
Putative transcription factor that appears to regulate lipid metabolism. [UniProt].
Specifications
ZFP69 | |
Polyclonal | |
Unconjugated | |
ZFP69 | |
Gm1029; Krab2; rCG30900-like; RP11-656D10.2; Zfp63; Zfp63p; Zfp69; ZFP69 zinc finger protein; ZFP69 zinc finger protein A; ZFP69A; Zfp965; zinc finger protein 642; Zinc finger protein 69; Zinc finger protein 69 homolog; zinc finger protein 69 homolog; zinc finger protein ZFP69; zinc finger protein 965; zinc finger protein ZFP69; ZKSCAN23A; ZNF642; ZSCAN54A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
339559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q49AA0 | |
ZFP69 | |
Recombinant protein corresponding to Human ZFP69. Recombinant protein control fragment (Product #RP-94526). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction