Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP91 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | ZFP91 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18013620
|
Novus Biologicals
NBP18013620UL |
20 μL |
Each for $152.22
|
|
NBP180136
|
Novus Biologicals
NBP180136 |
100 μL |
Each for $436.00
|
|
Description
ZFP91 Polyclonal specifically detects ZFP91 in Human samples. It is validated for Western Blot.Specifications
ZFP91 | |
Polyclonal | |
Rabbit | |
NP_444251 | |
80829 | |
Synthetic peptide directed towards the N terminal of human ZFP91. Peptide sequence PQAAKSPSPVQGKKSPRLLCIEKVTTDKDPKEEKEEEDDSALPQEVSIAA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
FLJ57065, ZFP-91, zinc finger protein 91 homolog (mouse), zinc finger protein homologous to Zfp91 in mouse | |
ZFP91 | |
IgG | |
Affinity Purified | |
63 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title