Learn More
Aviva Systems Biology Corp ZFPM1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32193_P050
This is a rabbit polyclonal antibody against ZFPM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFPM1. Alternate names: FOG, FOG1, ZNF408, ZNF89AApplicationWB. Species reactivity: Cow, Human, Rat
Catalog No. 50-014-54481