Learn More
Aviva Systems Biology Corp Zfy2 Antibody - middle region (ARP37006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfy2. It was validated on Western Blot. Zfy2,zinc finger protein 2, Y linked,Zfy-2; Zfy2,zinc finger Y-chromosomal protein 2,zinc finger Y-chromosomal protein 2,Y-linked zinc finger protein 2The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Zfy-2ApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-563
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.