Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFYVE1 Monoclonal Antibody (OTI4B3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527254
Description
ZFYVE1 Monoclonal antibody specifically detects ZFYVE1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ZFYVE1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HBF4 | |
ZFYVE1 | |
Human recombinant protein fragment corresponding to amino acids 1-308 of ZFYVE1 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4B3 | |
Unconjugated | |
ZFYVE1 | |
DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53349 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction