Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFYVE28 Mouse anti-Human, Clone: LST2/2426, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30715720UG
Description
ZFYVE28 Monoclonal specifically detects ZFYVE28 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
ZFYVE28 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array | |
hLst2, KIAA1643, lateral signaling target protein 2 homolog, LST2, LYST2, MGC43699, MGC61592, Zinc finger FYVE domain-containing protein 28, zinc finger, FYVE domain containing 28 | |
Recombinant full-length human ZFYVE28 protein | |
20 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Peptide Array | |
LST2/2426 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
57732 | |
Store at 4°C. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction