Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFYVE28 Mouse anti-Human, Clone: LST2/2426, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ZFYVE28
Clone LST2/2426
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121788
View Documents
Novus Biologicals
NBP30715720UG
20 μg
Each for $138.37
Add to cart
 
NB121787
View Documents
Novus Biologicals
NBP30715710UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ZFYVE28 Monoclonal antibody specifically detects ZFYVE28 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

ZFYVE28
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
57732
IgG2a Kappa
Primary
Store at 4°C.
LST2/2426
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array
Unconjugated
Mouse
Human
hLst2, KIAA1643, lateral signaling target protein 2 homolog, LST2, LYST2, MGC43699, MGC61592, Zinc finger FYVE domain-containing protein 28, zinc finger, FYVE domain containing 28
Recombinant full-length human ZFYVE28 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit