Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFYVE9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566804
Description
Immunogen sequence: NHLCPTSSDSL ASVCSPSQLK DDGSIGRDPS MSAITSLTVD SVISSQGTDG CPAVKKQENY IPDEDLTGKI SSPRTDLGSP N Highest antigen sequence identity to the following orthologs - mouse 62%, rat 60%.
Early endosomal protein that functions to recruit SMAD2/SMAD3 to intracellular membranes and to the TGF-beta receptor. Plays a significant role in TGF-mediated signaling by regulating the subcellular location of SMAD2 and SMAD3 and modulating the transcriptional activity of the SMAD3/SMAD4 complex. Possibly associated with TGF-beta receptor internalization.
Specifications
ZFYVE9 | |
Polyclonal | |
Unconjugated | |
Zfyve9 | |
E030027L17; hSARA; MAD, mothers against decapentaplegic homolog interacting protein, receptor activation anchor; MADH-interacting protein; MADHIP; Mothers against decapentaplegic homolog-interacting protein; Novel serine protease; NSP; PPP1R173; protein phosphatase 1, regulatory subunit 173; Receptor activation anchor; SARA; Smad anchor for receptor activation; SMADIP; ZFYVE9; Zinc finger FYVE domain-containing protein 9; zinc finger FYVE-type containing 9; zinc finger, FYVE domain containing 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9372 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95405 | |
Zfyve9 | |
Recombinant Human ZFYVE9. Recombinant protein control fragment (Product #RP-107461). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction