Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFYVE9 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ZFYVE9
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096032
View Documents
Novus Biologicals
NBP304604100UL
100 μg Each for $320.71
Add to cart
 
NB096033
View Documents
Novus Biologicals
NBP30460420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ZFYVE9 Polyclonal antibody specifically detects ZFYVE9 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

ZFYVE9
Unconjugated
Rabbit
Signal Transduction
PBS with 50% glycerol, pH7.3.
9372
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry
Purified
RUO
Human
Madh-interacting protein, MADHIP, mothers against decapentaplegic homolog interacting protein, mothers against decapentaplegic homolog interacting protein, receptoractivation anchor, NSP, receptor activation anchor, SARAhSARA, Smad anchor for receptor activation, SMADIPMAD, mothers against decapentaplegic homolog (Drosophila) interacting protein, zinc finger FYVE domain-containing protein 9, zinc finger, FYVE domain containing 9
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ZFYVE9 (NP_004790.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit