Learn More
ZG16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 173971AP
Catalog No. 50-173-1550
Add to cart
Specifications
Specifications
ZG16
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60844, Q8CJD3, Q8K0C5
hZG16, JCLN, JCLN1, Secretory lectin ZG16, ZG16, ZG16A, Zymogen granule protein 16
Rabbit
IgG
150 μL
-20°C
Primary
171449, 653808, 69036
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZG16
Liquid
Zg16
ZG16 Fusion Protein Ag11332
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat