Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZG16B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121494
This item is not returnable.
View return policy
Description
ZG16B Polyclonal antibody specifically detects ZG16B in Human samples. It is validated for Western Blot.
Specifications
| ZG16B | |
| Polyclonal | |
| Unconjugated | |
| ZG16B | |
| EECP; endocrine and exocrine protein; HRPE773; jacalin-like lectin domain containing 2; JCLN2; pancreatic adenocarcinoma upregulated factor; PAUF; PRO1567; UNQ773/PRO1567; ZG16B; Zymogen granule protein 16 homolog B; zymogen granule protein 16B | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 110259942, 124220 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96DA0 | |
| ZG16B | |
| Recombinant Zymogen Granule Protein 16 Homolog B (ZG16B), Gly17-Arg172 (Accession Q96DA0). | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction