Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZGPAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566034
Description
Immunogen sequence: ALCPSLAVVGS DAVDSGTCSS AFAGWEVHTR GIGSRLLTKM GYEFGKGLGR HAEGRVEPIH AVVLPRGKSL DQCVETLQKQ TRVG Highest antigen sequence identity to the following orthologs - mouse 78%, rat 78%.
Transcription repressor that specifically binds the 5'-GGAG[GA]A[GA]A-3' consensus sequence. Represses transcription by recruiting the chromatin multiprotein complex NuRD to target promoters. Negatively regulates expression of EGFR, a gene involved in cell proliferation, survival and migration. Its ability to repress genes of the EGFR pathway suggest it may act as a tumor suppressor. Able to suppress breast carcinogenesis. [UniProt].
Specifications
ZGPAT | |
Polyclonal | |
Unconjugated | |
ZGPAT | |
1500006I01Rik; BC021513; G patch domain-containing protein 6; GPATC6; GPATCH6; KIAA1847; RGD1310801; ZC3H9; ZC3HDC9; ZGPAT; Zinc finger and G patch domain-containing protein; zinc finger CCCH domain-containing protein 9; zinc finger CCCH-type and G-patch domain containing; zinc finger CCCH-type with G patch domain; zinc finger CCCH-type with G patch domain-containing protein; zinc finger, CCCH-type with G patch domain; zinc finger, CCCH-type with G patch domain isoform c; zinc finger, CCCH-type with G-patch domain; ZIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84619 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N5A5 | |
ZGPAT | |
Recombinant Human ZGPAT. Recombinant protein control fragment (Product #RP-105049). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction