Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZGPAT Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | ZGPAT |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15693720
|
Novus Biologicals
NBP15693720UL |
20 μL |
Each for $152.22
|
|
NBP156937
|
Novus Biologicals
NBP156937 |
100 μL |
Each for $436.00
|
|
Description
ZGPAT Polyclonal specifically detects ZGPAT in Human samples. It is validated for Western Blot.Specifications
ZGPAT | |
Polyclonal | |
Rabbit | |
Human | |
GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein | |
ZGPAT | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q8N5A5 | |
84619 | |
Synthetic peptides corresponding to ZGPAT(zinc finger, CCCH-type with G patch domain) The peptide sequence was selected from the middle region of ZGPAT. Peptide sequence LLREAVVEGDGILPPLRTEATESDSDSDGTGDSSYARVVGSDAVDSAQSS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title