Learn More
Abnova™ ZHX1 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00011244T01
Catalog No. 89-967-897
Add to cart
Description
Description
Specifications
Specifications
Western Blot
ZHX1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 96.14kDa
Transient overexpression cell lysate was tested with anti-ZHX1 antibody (Mfr. No. H00011244-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
NM_001017926.1
pCMV-ZHX1 full-length
100 μL
Human