Learn More
Aviva Systems Biology Corp ZHX3 Antibody - middle region (ARP57983_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57983_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZHX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZHX3. Alternate names: KIAA0395, TIX1ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-62201