Learn More
Selleck Chemical LLCSupplier Diversity Partner Zibotentan (ZD4054) 50mg 186497-07-4  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S145650MG
Zibotentan (ZD4054) is a specific Endothelin (ET)A antagonist with IC50 of 21 nM, exhibiting no activity at ETB. Phase 3.
Catalog No. 50-720-2