Learn More
Selleck Chemical LLC
Zibotentan (ZD4054) 10mg 186497-07-4
Manufacturer: Selleck Chemical LLC S145610MG
Zibotentan (ZD4054) is a specific Endothelin (ET)A antagonist with IC50 of 21 nM, exhibiting no activity at ETB. Phase 3.