Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIC2 Polyclonal Antibody
Supplier: Thermo Scientific PA5113616
Description

Specifications
ZIC2 | |
Polyclonal | |
Unconjugated | |
ZIC2 | |
cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 2; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; zinc finger protein ZIC 2; zinc finger protein Zic2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7546 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95409 | |
ZIC2 | |
Synthetic peptide directed towards the C terminal region of human ZIC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction