Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC3 Mouse anti-Human, Clone: OTI1G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274925

Catalog No. NBP274925


Add to cart

Description

Description

ZIC3 Monoclonal antibody specifically detects ZIC3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZIC3
OTI1G7
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
heterotaxy 1, HTXHTX1, Zic family member 3 (odd-paired homolog, Drosophila), Zinc finger protein 203, zinc finger protein ZIC 3, ZNF203Zinc finger protein of the cerebellum 3
Mouse
IgG1
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
7547
Human
Western Blot
LYOPH
Western Blot 1:2000
Affinity Purified, Lyophilized
ZIC3
Human recombinant protein fragment corresponding to amino acids 242-467 of human ZIC3(NP_003404) produced in E.coli.
50.4 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit