Learn More
ZIC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 217171AP
Catalog No. 50-173-3561
Add to cart
Description
Description
ZIC3 is a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in the gene encoding ZIC3 cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.
Specifications
Specifications
ZIC3
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60481, Q62521
HTX, HTX1, ZIC3, Zinc finger protein ZIC 3, ZNF203
Rabbit
IgG
150 μL
-20°C
Primary
22773, 7547
Western Blot
Unconjugated
ZIC3
Liquid
ZIC3
ZIC3 Fusion Protein Ag16354
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse