ZICO™ Load and Lock™ Walkaway™ Bracket
Secure, positive latching mechanism meets NFPA 1901-09 requirements for mounting SCBAs in driving or crew compartments
Manufacturer: Ziamatic CorporationSupplier Diversity Partner 1054012000
 View more versions of this product
Catalog No. 19-123-289
  Check Availability
Add to Cart
    Specifications
    Product Type Assembled Version Bracket
    For Use With (Application) Storage of SCBAs in crew or driving compartments
    Includes Straps
    Weight (Metric) 2.4kg
    Weight (English) 5.4 lb.
    Certifications NFPA 1901-09
  • Description & Specifications
Related Products
Product Content Feedback