Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika virus envelope protein Mouse anti-Virus, Clone: M86648, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14217

Catalog No. 89-033-717


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant Zika virus envelope protein.

  • High specificity
  • High reactivity
Specifications

Specifications

Zika virus envelope protein
M86648
Mouse monoclonal antibody raised against partial recombinant Zika virus envelope protein.
In 0.015 M potassium phosphate, 0.85% NaCl, pH 7.2 (0.05% sodium azide).
Mouse
IgG1, kappa
500 ug
Store at 4°C.
Monoclonal
Virus
ELISA, Lateral Flow
Unconjugated
ELISA Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to amino acids 291-690 of Zika virus envelope protein.
DEAE column chromatography
RUO
Primary
No cross reactivity with recombinant Dengue and Chikungunya antigens.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)