Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Zika Virus Envelope (Q32Ze1, 291 A.A.-690 A.A.) Partial Recombinant Protein Expressed In Baculovirus Expression System

Manufacturer:  Abnova™ P6207

Catalog No. 89-033-712


Add to cart

Description

Description

High quality
Specifications

Specifications

Q32ZE1
Liquid
7751225
Baculovirus expression system
100 ug
RUO
Enzyme-linked Immunoabsorbent Assay, Lateral-Flow Immunochromatographic Assay, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
Zika virus Envelope Recombinant Protein
Coomassie stained SDS-PAGE gel
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Insect
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only