Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus NS3 Rabbit anti-Virus, FITC, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc ZIKANS3FITC

Description
Zika Virus NS3 Polyclonal antibody specifically detects Zika Virus NS3 in Virus samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
Zika Virus NS3 | |
Polyclonal | |
FITC | |
NS3 protease; Zika NS3 protease; Zika Virus NS3 protease | |
Synthetic peptide taken within amino acid region 1-50 on Zika virus non structural protein NS3. | |
200 μL | |
Primary | |
-20° C, store in dark | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title