Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus Protein 2K Rabbit anti-Virus, Biotin, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc ZIKAP2KBIOTIN

Description
Zika Virus Protein 2K Polyclonal antibody specifically detects Zika Virus Protein 2K in Virus samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
Zika Virus Protein 2K | |
Polyclonal | |
Biotin | |
Zika Protein 2K; Zika proteolytic helicase; ZIKA2PK | |
23 mer Synthetic peptide taken from aa 2247 through 2269 of the ZIKA polyprotein AHL43505. | |
200 μL | |
Primary | |
-20°C | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title