Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus VLP Mouse anti-Virus, Clone: IG3-1-A12-G9, Novus Biologicals™

Mouse Monoclonal Antibody
$515.50 - $1262.00
Specifications
Antigen | Zika Virus VLP |
---|---|
Clone | IG3-1-A12-G9 |
Concentration | 1 mg/mL |
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Description
Zika Virus VLP Monoclonal antibody specifically detects Zika Virus VLP in Virus samples. It is validated for Western Blot, ELISASpecifications
Zika Virus VLP | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Virus | |
polyprotein, partial | |
IgG1 κ | |
Protein G purified | |
Antibody |
IG3-1-A12-G9 | |
Western Blot, ELISA | |
Monoclonal | |
Purified | |
RUO | |
PBS pH 7.4 | |
Recombinant Zika virus VLPs (comprising envelope, pre-membrane and membrane proteins), produced in HEK293 cells | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title