Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIM2 Mouse anti-Human, Clone: OTI6D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274928

Catalog No. NBP274928


Add to cart

Description

Description

ZIM2 Monoclonal antibody specifically detects ZIM2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZIM2
OTI6D12
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
zinc finger imprinted 2, zinc finger, imprinted 2, ZNF656Zinc finger protein 656
Mouse
IgG1
Immunogen affinity purified
modENCODE Antibodies
Primary
23619
Human
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:500
Affinity Purified, Lyophilized
ZIM2
Human recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 (NP_056178) produced in E.coli.
61 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit