Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIM2 Mouse anti-Human, PE, Clone: OTI7G1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274929PE

Catalog No. NBP274929PE


Add to cart

Description

Description

ZIM2 Monoclonal antibody specifically detects ZIM2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ZIM2
OTI7G1
Flow Cytometry
Affinity Purified, Conjugated
ZIM2
Recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 (NP_056178) produced in E.coli.
Immunogen affinity purified
modENCODE Antibodies
Primary
23619.0
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
zinc finger imprinted 2, zinc finger, imprinted 2, ZNF656Zinc finger protein 656
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit