Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZIM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556979
Description
Immunogen sequence: NYSNLVSVGQ GETTKPDVIL RLEQGKEPWL EEEEVLGSGR AEKNGDIGGQ IWKPKDVKES LAREVPSINK ETLTTQKGVE CDGSKK Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 35%.
ZIM3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 11 C2H2-type zinc fingers and 1 KRAB domain. ZIM3 may be involved in transcriptional regulation.
Specifications
ZIM3 | |
Polyclonal | |
Unconjugated | |
ZIM3 | |
ZIM3; Zinc finger imprinted 3; zinc finger protein 657; zinc finger, imprinted 3; ZNF657 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PE6 | |
ZIM3 | |
Recombinant protein corresponding to Human ZIM3. Recombinant protein control fragment (Product #RP-95514). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction