Alfa Aesar™ Zinc 2,4-pentanedionate monohydrate

Manufacturer: Alfa Aesar™ 1252718

Quantity 50g

 View more versions of this product

Catalog No. AA1252718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14363-15-6
    Formula Weight 281.60 (263.59 Anhydrous)
    MDL Number MFCD00000035
    Synonym Zinc acetylacetonate; Bis(acetylacetonato)zinc(II) monohydrate
    Chemical Name or Material Zinc 2,4-pentanedionate monohydrate

    Specifications

    Formula Weight 281.60 (263.59 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 136° to 138°C (dec.)
    MDL Number MFCD00000035
    Solubility Information Soluble in alcohol; Slightly soluble in water
    Health Hazard 1 Warning
    Physical Form Crystalline
    CAS 14363-15-6
    Density 1.6g/mL
    TSCA TSCA
    Synonym Zinc acetylacetonate; Bis(acetylacetonato)zinc(II) monohydrate
    Quantity 50g
    Chemical Name or Material Zinc 2,4-pentanedionate monohydrate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit