Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc, 5% suspension in THF, highly active Rieke™ metal, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
7440-66-6
Zn
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
7440-66-6
Zn
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
Flash Point −17°C
Specific Gravity 0.949
Quantity 100mL
Density 0.9490g/mL
Linear Formula Zn
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Packaging Glass bottle
Formula Weight 65.38
Chemical Name or Material Zinc, 5% suspension in THF, highly active Rieke™ metal

GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
May form explosive peroxides.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Handle under inert gas. Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.