Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc, 99.999% (trace metal basis), powder, -40 mesh, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust,cinc,powder,merrillite,rheinzink,jasad,blue powder,granular,zinc, elemental,emanay dust
23994
CHEBI:30185
zinc
[Zn]
for ultra trace analysis, Reduction reagent
Quantity 10g
Density 7.14
Assay Percent Range 100.00%
Linear Formula Zn
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Merck Index 15, 10326
Packaging Glass bottle
Formula Weight 65.38
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Total Trace Metal Impurities 0.001% max.
Color Gray
Melting Point 420.0°C
Boiling Point 907.0°C
Chemical Name or Material Zinc, 99.999%
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.
Catches fire spontaneously if exposed to air.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger