Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc, +99%, mossy, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]
Reduction reagent
Specific Gravity 7.14
Quantity 1kg
Density 7.1400g/mL
Assay Percent Range 99+%
Linear Formula Zn
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Merck Index 15, 10326
Packaging Plastic bottle
Formula Weight 65.38
Physical Form Flakes
Percent Purity 99+%
Total Trace Metal Impurities 1.0% max.
Color Gray
Melting Point 419.0°C
Boiling Point 907.0°C
Chemical Name or Material Zinc, 99+%
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-175-3

RTECSNumber : ZG8600000

TSCA : TSCA