Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Acetate, Crystal, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
5970-45-6
C4H10O6Zn
219.50
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc(2+) diacetate dihydrate
O.O.[Zn++].CC([O-])=O.CC([O-])=O
5970-45-6
C4H10O6Zn
219.50
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc(2+) diacetate dihydrate
O.O.[Zn++].CC([O-])=O.CC([O-])=O

ZI200, 5970-45-6, C4H6O4Zn•2H2O

Spectrum™ Zinc Acetate, Crystal, USP is used in anti-itch oitments and for treating zinc deficiencies. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 98 to 102%
Insoluble Matter 5e-005
Lead (Pb) 2e-005
Packaging Fiber Drum
Sulfate 0.0001
pH 6.0 to 8.0
Quantity 50 kg