Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Acetate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

5970-45-6
C4H10O6Zn
219.498
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
2724192

High quality chemicals for laboratory and specialized industrial use

SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
IUPAC Name zinc;diacetate;dihydrate
Formula Weight 219.51
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.74kg
Quantity 2.5kg
Chemical Name or Material Zinc Acetate, Dihydrate
Grade Baker Analyzed™ ACS Reagent