Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc acetate dihydrate, Honeywell™

Click to view available options
Quantity:
250g
5970-45-6
C4H10O6Zn
219.498
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate,zinc diacetate dihydrate,zinc ii acetate dihydrate,unii-fm5526k07a,acetic acid zinc salt,zinc acetate usp,acetic acid, zinc salt, dihydrate,zinc acetate, dihydrate,galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
5970-45-6
C4H10O6Zn
219.498
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate,zinc diacetate dihydrate,zinc ii acetate dihydrate,unii-fm5526k07a,acetic acid zinc salt,zinc acetate usp,acetic acid, zinc salt, dihydrate,zinc acetate, dihydrate,galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Tested according to European Pharmacopoeia

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3732513
Formula Weight 219.51g/mol
Percent Purity 99 to 101%
Quantity 250g
Chemical Name or Material Zinc acetate dihydrate

P273-P305 + P351 + P338-P501

RTECSNumber : ZG8750000