Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zinc acetylacetonate hydrate, 96%

Click to view available options
Quantity:
100g
500g
108503-47-5
C10H16O5Zn
281.613
MFCD00149060
KUJHAYOLESEVSA-SUKNRPLKSA-L
zinc acetylacetonate hydrate,zinc 2,4-pentanedionate, hydrate,zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
131675103
zinc;(Z)-4-oxopent-2-en-2-olate;hydrate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Zn+2]
108503-47-5
C10H16O5Zn
281.613
MFCD00149060
KUJHAYOLESEVSA-SUKNRPLKSA-L
zinc acetylacetonate hydrate,zinc 2,4-pentanedionate, hydrate,zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
131675103
zinc;(Z)-4-oxopent-2-en-2-olate;hydrate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Zn+2]
Quantity 500g
Melting Point ≥137°C (decomposition)
Formula Weight 263.59
Percent Purity 96%
Chemical Name or Material Zinc acetylacetonate hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : ZH0917500

TSCA : TSCA