Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Zinc arsenide, 99.999% (metals basis)
Click to view available options
Quantity:
2g
10g
Specifications
UN Number | UN1557 |
Solubility Information | It is insoluble in water. |
Formula Weight | 215.21 |
Physical Form | 10mm & Down Lump |
Percent Purity | 99.999% |
Odor | Odorless |
Particle Size | 10mm & down |
Quantity | 10g |
Chemical Name or Material | Zinc arsenide |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 234-956-7
TSCA : Yes
Recommended Storage : Ambient temperatures