Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050215
Click to view available options
Quantity:
10 g
12044-55-2
As2Zn
215.223
MFCD00151429
KXUSEUKXCIKTBF-UHFFFAOYSA-N
zinc arsenide znas2, 1,2-diarsa-3-zincacycloprop-1-ene
102601572
$l^{2}-arsanylidenearsenic;zinc
[Zn].[As]=[As]
12044-55-2
As2Zn
215.223
MFCD00151429
KXUSEUKXCIKTBF-UHFFFAOYSA-N
zinc arsenide znas2, 1,2-diarsa-3-zincacycloprop-1-ene
102601572
$l^{2}-arsanylidenearsenic;zinc
[Zn].[As]=[As]

Zinc arsenide is often used as a component in the manufacture of semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Lump
Quantity 10 g
UN Number UN1557
Solubility Information It is insoluble in water.
Formula Weight 215.21
Percent Purity 99.999%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc arsenide
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 234-956-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit