Alfa Aesar™ Zinc bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 4688003

Quantity 1g

 View more versions of this product

Catalog No. AA4688003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 168106-25-0
    Molecular Formula C4F12N2O8S4Zn
    Linear Formula [(CF3SO2)2N]2Zn
    Formula Weight 625.65
    MDL Number MFCD16621474
    Synonym Zinc triflimide; Zinc trifluoromethanesulfonimide
    Chemical Name or Material Zinc bis (trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 625.65
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD16621474
    Quantity 1g
    Sensitivity Moisture Sensitive
    CAS 168106-25-0
    Health Hazard 1 Warning
    Color White
    Melting Point 143° to 147°C
    Physical Form Solid
    Linear Formula [(CF3SO2)2N]2Zn
    Synonym Zinc triflimide; Zinc trifluoromethanesulfonimide
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C4F12N2O8S4Zn
    Flash Point >110°C (230°F)
    Chemical Name or Material Zinc bis (trifluoromethylsulfonyl)imide

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit