Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc bromide, 99.999%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass bottle
7699-45-8
Br2Zn
225.19
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
7699-45-8
Br2Zn
225.19
MFCD00011294
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula ZnBr2
Fieser 02,463; 05,762; 08,535; 09,520; 10,461; 11,600; 13,349; 15,368; 16,389
Merck Index 15, 10328
Solubility Information Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether,solutions of alkali hydroxides,1g/0.5mL 90% alcohol
Formula Weight 225.19
Physical Form Crystalline Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Density 4.2200g/mL
Packaging Glass bottle
Solubility Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether, solutions of alkali hydroxides, 1g/0.5mL 90% alcohol
Specific Gravity 4.22
Total Trace Metal Impurities 0.001% max
Boiling Point 697.0°C
Color White
Melting Point 394.0°C
Quantity 2.5g
Chemical Name or Material Zinc bromide, 99.999%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-718-4

RTECSNumber : ZH1150000

TSCA : TSCA