Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc bromide, 98%, extra pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
7699-45-8
Br2Zn
225.19
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
7699-45-8
Br2Zn
225.19
VNDYJBBGRKZCSX-UHFFFAOYSA-L
zincbromide,zinc ii bromide,acmc-209p7h,zinc 2+ ion dibromide,ksc378m6n,zinc bromide hydrate, puratronic™,zinc bromide, anhydrous 100g
10421208
zinc;dibromide
[Zn+2].[Br-].[Br-]
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula ZnBr2
Fieser 02,463; 05,762; 08,535; 09,520; 10,461; 11,600; 13,349; 15,368; 16,389
Merck Index 15,10328
Solubility Information Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether,solutions of alkali hydroxides,1g/0.5mL 90% alcohol
Barium (Ba) 10ppm max.
Formula Weight 225.19
Physical Form Powder and/or Chunks
Percent Purity 98%
Grade Extra Pure
Calcium (Ca) 100ppm max.
Density 4.2200g/mL
Iron (Fe) 10ppm max.
Lead (Pb) 50ppm max.
Magnesium (Mg) 100ppm max.
Packaging Glass bottle
Potassium (K) 100ppm max.
Sodium (Na) 100ppm max.
Solubility Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether, solutions of alkali hydroxides, 1g/0.5mL 90% alcohol
Specific Gravity 4.22
Sulfate 100ppm max.
Boiling Point 697.0°C
Color White
Melting Point 394.0°C
Quantity 2.5kg
Chemical Name or Material Zinc bromide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-718-4