Alfa Aesar™ Zinc bromide, anhydrous, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4422309

Quantity 10g

 View more versions of this product

Catalog No. AA4422309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7699-45-8
    Molecular Formula ZnBr2
    Formula Weight 225.19
    MDL Number MFCD00011294
    UN Number UN3260
    Merck Index 14,10129
    Chemical Name or Material Zinc bromide, Anhydrous

    Specifications

    Formula Weight 225.19
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Refractive Index 1.5452
    MDL Number MFCD00011294
    CAS 7699-45-8
    Boiling Point 650°C
    Color White
    Quantity 10g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Storage Note 1
    • Packaged under Argon
    Sensitivity Hygroscopic
    Merck Index 14,10129
    Density 4.201g/mL
    Physical Form Lumps
    Molecular Formula ZnBr2
    UN Number UN3260
    Melting Point 394°C
    RTECS Number ZH1150000
    TSCA TSCA
    Chemical Name or Material Zinc bromide, Anhydrous
    Assay 99.999%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit