Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc Bromide, Anhydrous, Reagent, 98%, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
100 g
500 g
6 x 500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
Description
Spectrum™ Chemical Zinc Bromide, Anhydrous, Reagent is a salt that has no color and share many similarities with zinc chloride. This inorganic compound is very soluble in water and is used as a Lewis acid and in an aqueous solution as a transparent shield against radiation in between two glass panes. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications
CAS Max % | 1 |
Quantity | 100 g |
Percent Purity | 98% |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction